Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_213629.1 | NP_998794.1 | 15 | 2229 | ||
XM_006243047.2 | XP_006243109.1 | 15 | 2240 | |||
GenBank mRNA | AY501475.1 | - | 15 | 2229 |