Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acy3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009603.1 | NP_001009603.1 | 7 | 969 | ||
XM_006230733.2 | XP_006230795.1 | 7 | 978 | |||
XM_006230734.2 | XP_006230796.1 | 7 | 1646 | |||
XM_008760098.1 | XP_008758320.1 | 7 | 1139 | |||
XM_008760099.1 | XP_008758321.1 | 7 | 1001 | |||
GenBank mRNA | BC088190.1 | - | 7 | 969 | 7371632 |