Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfsf13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009623.1 | NP_001009623.1 | 6 | 1375 | ||
XM_006246625.2 | XP_006246687.1 | 8 | 1722 | |||
XM_008767796.1 | XP_008766018.1 | 7 | 1878 | |||
XM_008767797.1 | XP_008766019.1 | 7 | 1875 | |||
XM_008767798.1 | XP_008766020.1 | 6 | 1830 | |||
XM_008767799.1 | XP_008766021.1 | 6 | 1827 | |||
XM_008767800.1 | XP_008766022.1 | 6 | 1784 | |||
XM_008767801.1 | XP_008766023.1 | 6 | 1781 | |||
XM_008767802.1 | XP_008766024.1 | 5 | 2068 | |||
GenBank mRNA | BC087135.1 | - | 6 | 1375 | 7123934 | |
FQ215356.1 | - | 6 | 813 |