Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apoh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009626.1 | NP_001009626.1 | 2, 3 | 243 | ||
GenBank mRNA | AM071386.1 | - | 2, 3 | 202 | ||
BC088180.1 | - | 2, 3 | 243 | 7364928 | ||
FQ209412.1 | - | 2, 3 | 282 | |||
FQ209491.1 | - | 2, 3 | 269 | |||
FQ209740.1 | - | 2, 3 | 278 | |||
FQ210182.1 | - | 2, 3 | 269 | |||
FQ210311.1 | - | 2, 3 | 269 | |||
FQ210756.1 | - | 2, 3 | 285 | |||
FQ210813.1 | - | 2, 3 | 269 | |||
FQ211328.1 | - | 2, 3 | 242 | |||
FQ218445.1 | - | 2, 3 | 283 | |||
FQ218454.1 | - | 2, 3 | 269 | |||
FQ218597.1 | - | 2, 3 | 265 | |||
FQ218599.1 | - | 2, 3 | 269 | |||
FQ218637.1 | - | 2, 3 | 269 | |||
FQ218785.1 | - | 2, 3 | 285 | |||
FQ218859.1 | - | 2, 3 | 242 | |||
FQ218865.1 | - | 2, 3 | 269 | |||
FQ218961.1 | - | 2, 3 | 269 | |||
FQ219138.1 | - | 2, 3 | 334 | |||
FQ219328.1 | - | 2, 3 | 269 | |||
FQ219380.1 | - | 2, 3 | 270 | |||
FQ219438.1 | - | 2, 3 | 268 | |||
FQ219481.1 | - | 2, 3 | 282 | |||
FQ219537.1 | - | 2, 3 | 266 | |||
FQ219580.1 | - | 2, 3 | 283 | |||
FQ219601.1 | - | 2, 3 | 282 | |||
FQ219674.1 | - | 2, 3 | 269 |