Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193335.1 | NP_001180264.1 | 2 | 404 | ||
NM_017680.4 | NP_060150.4 | 2 | 404 | |||
GenBank mRNA | AF316824.1 | - | 1 | 122 | ||
AK027359.1 | - | 2 | 195 | |||
AK172737.1 | - | 2 | 221 | |||
AK304501.1 | - | 2 | 214 | |||
AK308107.1 | - | 2 | 317 | |||
AY029191.1 | - | 2 | 331 | |||
AY358329.1 | - | 2 | 337 | |||
BC022059.1 | - | 2 | 352 | |||
BC063114.1 | - | 2 | 354 | 30404602 | ||
BP292032.1 | - | 2 | 404 | |||
CD110559.1 | - | 351 |