Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009637.1 | NP_001009637.1 | 9 | 938 | ||
XM_006254672.2 | XP_006254734.1 | 9 | 1044 | |||
GenBank mRNA | BC087655.1 | - | 9 | 938 | 7312531 | |
FQ212494.1 | - | 9 | 975 |