Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009645.1 | NP_001009645.1 | 14 | 2009 | ||
GenBank mRNA | BC088421.1 | - | 14 | 2009 | 7123493 |