Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009647.1 | NP_001009647.1 | 3 | 280 | ||
GenBank mRNA | BC058154.1 | - | 3 | 284 | ||
BC088312.1 | - | 3 | 280 | 7324682 | ||
EU148065.1 | - | 2 | 111 | |||
FQ212609.1 | - | 3 | 290 | |||
FQ214728.1 | - | 1 | 68 | |||
FQ215871.1 | - | 3 | 292 | |||
FQ219747.1 | - | 3 | 278 |