Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp622 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009652.1 | NP_001009652.1 | 1228 | |||
GenBank mRNA | BC088214.1 | - | 1228 | 7383802 |
Gene Symbol: Fam134b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY310142.1 | - | 2258 | |||
AY321329.1 | - | 2253 |
Gene Symbol: LOC103690028 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760822.1 | XP_008759044.1 | 4 | 2366 |