Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zgpat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009656.1 | NP_001009656.1 | 1 | 386 | ||
XM_006235749.1 | XP_006235811.1 | 2 | 382 | |||
XM_006235750.1 | XP_006235812.1 | 2 | 347 | |||
XM_006235751.2 | XP_006235813.1 | 2 | 368 | |||
XM_006235752.2 | XP_006235814.1 | 1 | 282 | |||
GenBank mRNA | BC087720.1 | - | 1 | 386 | 7318724 |