Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009657.1 | NP_001009657.1 | 8, 9 | 869 | ||
GenBank mRNA | BC087663.1 | - | 8, 9 | 869 | 7315259 |