Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009662.1 | NP_001009662.1 | 2 | 350 | ||
GenBank mRNA | BC087586.1 | - | 2 | 350 | 7318288 |