Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM102B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010883.2 | NP_001010883.2 | 11 | 4007 | ||
GenBank mRNA | AK025203.1 | - | 1 | 470 | ||
AK094905.1 | - | 1 | 1048 | |||
BC140863.1 | - | 11 | 3863 | 9021731 | ||
BC140865.1 | - | 11 | 3863 | 9021733 | ||
CR749397.1 | - | 8 | 3577 | |||
CR936815.1 | - | 4 | 6990 |