Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242729.1 | NP_001229658.1 | 1 | 222 | ||
NM_017700.1 | NP_060170.1 | 1 | 222 | |||
XM_011532051.2 | XP_011530353.1 | 1 | 237 | |||
GenBank mRNA | AK000191.1 | - | 1 | 222 | ||
BC105695.1 | - | 1 | 210 | 40023595 |