Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr1500 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011831.1 | NP_001011831.1 | 1 | 138 | ||
GenBank mRNA | BC150926.1 | - | 1 | 203 | 9087837 |