Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011892.1 | NP_001011892.1 | 7 | 619 | ||
XM_006246675.1 | XP_006246737.1 | 7 | 764 | |||
GenBank mRNA | BC079362.1 | - | 7 | 619 | 7109243 |