Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011904.1 | NP_001011904.1 | 5 | 1232 | ||
XM_006249434.2 | XP_006249496.1 | 5 | 809 | |||
GenBank mRNA | BC083831.1 | - | 5 | 1232 | 7112480 |