Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011914.1 | NP_001011914.1 | 3 | 2383 | ||
GenBank mRNA | BC083822.1 | - | 3 | 2383 | 7111461 |
Gene Symbol: LOC100910207 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751317.3 | XP_003751365.1 | 2472 |