Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anxa11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011918.1 | NP_001011918.1 | 15 | 1788 | ||
XM_008770980.1 | XP_008769202.1 | 15 | 2034 | |||
GenBank mRNA | BC083812.1 | - | 15 | 1788 | 7110191 |