Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtpbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011919.1 | NP_001011919.1 | 9 | 2287 | ||
GenBank mRNA | BC087083.1 | - | 9 | 2287 | 7189218 |