Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011950.2 | NP_001011950.2 | 5 | 672 | ||
XM_006232610.2 | XP_006232672.1 | 5 | 468 | |||
XM_006232611.2 | XP_006232673.1 | 5 | 468 | |||
GenBank mRNA | BC085878.1 | - | 5 | 672 | 7191747 | |
FQ230674.1 | - | 5 | 462 |