Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011953.1 | NP_001011953.1 | 4, 5 | 386 | ||
XM_008761549.1 | XP_008759771.1 | 4, 5 | 412 | |||
GenBank mRNA | BC083636.1 | - | 4, 5 | 386 | 7134753 |