Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cybrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011954.1 | NP_001011954.1 | 4 | 1164 | ||
XM_008761891.1 | XP_008760113.1 | 3 | 1312 | |||
GenBank mRNA | BC085768.1 | - | 4 | 1164 | 7108043 |