Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083590.1 | NP_001077059.1 | 7 | 937 | ||
NM_017727.4 | NP_060197.4 | 8 | 1072 | |||
XM_005264381.3 | XP_005264438.1 | 8 | 1045 | |||
XM_005264382.3 | XP_005264439.1 | 8 | 1045 | |||
GenBank mRNA | AF289555.1 | - | 2 | 108 | ||
AK000261.1 | - | 3 | 189 | |||
AK074981.1 | - | 8 | 1000 | |||
AK097308.1 | - | 7 | 867 | |||
AK315359.1 | - | 6 | 536 | |||
AL833725.1 | - | 8 | 932 | |||
AL834157.1 | - | 3 | 178 | |||
BC068479.1 | - | 8 | 1024 | 4814193 |