Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cercam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011962.1 | NP_001011962.1 | 11, 12 | 1259 | ||
XM_006233768.2 | XP_006233830.1 | 8, 9 | 1927 | |||
XM_006233769.2 | XP_006233831.1 | 9, 10 | 1319 | |||
XM_006233770.2 | XP_006233832.1 | 11, 12 | 2299 | |||
XM_006233771.2 | XP_006233833.1 | 7, 8 | 1793 | |||
GenBank mRNA | BC085782.1 | - | 11, 12 | 1259 | 7107449 |