Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xkr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011971.1 | NP_001011971.1 | 4 | 2127 | ||
GenBank mRNA | AY534258.1 | - | 4 | 2127 |