Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp6v0d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011972.1 | NP_001011972.1 | 8 | 1116 | ||
GenBank mRNA | BC089917.1 | - | 8 | 1116 | 7306609 |