Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glipr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011987.1 | NP_001011987.1 | 2 | 349 | ||
GenBank mRNA | BC089858.1 | - | 2 | 349 | 7384037 | |
FQ219892.1 | - | 2 | 354 | |||
FQ223414.1 | - | 2 | 354 | |||
FQ226490.1 | - | 2 | 370 | |||
FQ227021.1 | - | 2 | 369 | |||
FQ229493.1 | - | 2 | 376 | |||
FQ229810.1 | - | 2 | 415 | |||
FQ230706.1 | - | 2 | 377 |