Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pycrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011993.1 | NP_001011993.1 | 4 | 467 | ||
GenBank mRNA | BC087166.1 | - | 4 | 467 | 7135468 |