Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gga1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011994.1 | NP_001011994.1 | 16 | 2703 | ||
XM_006241973.2 | XP_006242035.1 | 17 | 4065 | |||
XM_008765679.1 | XP_008763901.1 | 18 | 3821 | |||
XM_008765680.1 | XP_008763902.1 | 15 | 3319 | |||
XM_008765681.1 | XP_008763903.1 | 17 | 6527 | |||
GenBank mRNA | BC090031.1 | - | 16 | 2703 | 7366311 |