Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C2cd2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011996.1 | NP_001011996.1 | 14 | 3240 | ||
XM_006242917.2 | XP_006242979.1 | 14 | 3297 | |||
XM_006242918.2 | XP_006242980.1 | 14 | 3294 | |||
XM_006242919.2 | XP_006242981.1 | 14 | 3294 | |||
XM_008766157.1 | XP_008764379.1 | 14 | 2849 | |||
GenBank mRNA | BC085925.1 | - | 14 | 3240 | 7109537 |