Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011998.1 | NP_001011998.1 | 6 | 804 | ||
XM_006243419.2 | XP_006243481.1 | 6 | 522 | |||
GenBank mRNA | BC085831.1 | - | 6 | 804 | 7134220 |