Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012012.2 | NP_001012012.2 | 6 | 444 | ||
NM_001199169.1 | NP_001186098.1 | 5 | 384 | |||
XM_006247195.2 | XP_006247257.1 | 7 | 837 | |||
XM_006247196.2 | XP_006247258.1 | 6 | 614 | |||
XM_006247198.2 | XP_006247260.1 | 5 | 554 | |||
XM_008768047.1 | XP_008766269.1 | 7 | 1188 | |||
XM_008768048.1 | XP_008766270.1 | 7 | 1338 | |||
GenBank mRNA | BC086543.1 | - | 6 | 442 | 7132863 | |
DN932564.1 | - | 379 | ||||
FQ212083.1 | - | 7 | 498 |