Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161584.1 | NP_001155056.1 | 2 | 305 | ||
NM_017746.3 | NP_060216.2 | 2 | 361 | |||
XM_011518798.1 | XP_011517100.1 | 2 | 323 | |||
XM_017014848.1 | XP_016870337.1 | 2 | 323 | |||
GenBank mRNA | AB060968.1 | - | 2 | 277 | ||
AK000294.1 | - | 2 | 277 | |||
AK298916.1 | - | 2 | 280 | |||
AK302619.1 | - | 2 | 308 | |||
AK303662.1 | - | 2 | 274 | |||
AY598337.1 | - | 1 | 146 | |||
BC030652.2 | - | 2 | 197 | 5268105 | ||
BP336313.1 | - | 2 | 361 |