Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ftsj3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012014.1 | NP_001012014.1 | 14 | 1396 | ||
GenBank mRNA | BC086512.1 | - | 14 | 1396 | 7133710 |