Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc39a12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012305.2 | NP_001012305.1 | 7 | 1368 | ||
GenBank mRNA | AK134622.1 | - | 7 | 1368 | ||
BC089362.1 | - | 7 | 1362 | 30628112 | ||
BC113764.1 | - | 7 | 1351 | 40059402 | ||
BC113765.1 | - | 7 | 1351 | 40059404 |