Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CCDC66 SIRNA Assays ›
Gene Symbol:
CCDC66
Gene Name:
coiled-coil domain containing 66
Gene Aliases:
-
Chromosome Location:
Chr.3: 56557125 - 56621837 on Build GRCh38
UniGene ID:
Hs.476399
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
252313
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CCDC66
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001012506.4
NP_001012524.4
17
2800
NM_001141947.1
NP_001135419.1
17
2819
NR_024460.1
17
2800
XM_005265081.3
XP_005265138.1
17
2837
XM_005265082.3
XP_005265139.1
17
2816
XM_005265083.3
XP_005265140.1
17
2708
XM_005265084.3
XP_005265141.1
17
2831
XM_011533614.2
XP_011531916.1
16
2842
XM_011533615.2
XP_011531917.1
16
2852
XM_011533616.2
XP_011531918.1
17
2776
XM_011533617.2
XP_011531919.1
18
2934
XM_011533619.2
XP_011531921.1
18
2837
XM_017006229.1
XP_016861718.1
17
2798
XM_017006230.1
XP_016861719.1
16
2820
XM_017006231.1
XP_016861720.1
16
2823
XM_017006232.1
XP_016861721.1
17
2690
XM_017006233.1
XP_016861722.1
17
2669
XM_017006234.1
XP_016861723.1
17
2858
XM_017006235.1
XP_016861724.1
18
2916
XM_017006236.1
XP_016861725.1
19
2929
XM_017006237.1
XP_016861726.1
19
2966
XM_017006238.1
XP_016861727.1
20
3107
XM_017006239.1
XP_016861728.1
14
2167
XM_017006240.1
XP_016861729.1
18
2870
XM_017006241.1
XP_016861730.1
19
2829
XM_017006242.1
XP_016861731.1
18
2692
XM_017006243.1
XP_016861732.1
17
2785
XM_017006244.1
XP_016861733.1
18
2921
XM_017006245.1
XP_016861734.1
16
2624
GenBank mRNA
AK096754.1
-
4
2135
AL832556.1
-
2
2524
AL832692.1
-
17
2740
AL833326.1
-
18
2837
BC047509.1
-
17
2768
5276957
BC132827.1
-
17
2789
40146849
Back To Top
More Information
Gene Ontology Categories:
Function(s)
retinal rod cell development
detection of light stimulus involved in visual perception
post-embryonic retina morphogenesis in camera-type eye
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-vpmkj:80/100.66.78.144:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline