Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01588 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126497.1 | 4 | 1606 | |||
NR_126498.1 | 4 | 2233 | ||||
GenBank mRNA | AK091232.1 | - | 4 | 2232 |