Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLPPR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017753.2 | NP_060223.2 | 3 | 400 | ||
NM_207299.1 | NP_997182.1 | 3 | 533 | |||
GenBank mRNA | AK000307.1 | - | 3 | 400 | ||
AK225306.1 | - | 3 | 400 | |||
AK303718.1 | - | 1 | 280 | |||
AK303807.1 | - | 2 | 396 | |||
AK309115.1 | - | 1 | 109 | |||
AY304515.1 | - | 3 | 266 | |||
AY337718.1 | - | 2 | 146 | |||
BC022465.1 | - | 3 | 266 | 4797229 | ||
BI601921.1 | - | 3 | 539 | |||
BI914341.1 | - | 3 | 573 | |||
CA772610.1 | - | 3 | 436 |