Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ckmt1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012738.1 | NP_001012756.1 | 4 | 680 | ||
XM_006234830.2 | XP_006234892.1 | 5 | 636 | |||
GenBank mRNA | BC091335.1 | - | 4 | 680 | 7314893 | |
X59737.1 | - | 5 | 590 |