Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sectm1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013043.1 | NP_001013061.1 | 2, 3 | 229 | ||
XM_006247885.2 | XP_006247947.1 | 2, 3 | 256 | |||
GenBank mRNA | BC078937.1 | - | 2, 3 | 229 | 7108345 | |
FQ225276.1 | - | 2, 3 | 222 |