Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MBTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017643.2 | NP_060113.2 | 8 | 983 | ||
XM_005257467.4 | XP_005257524.1 | 7 | 928 | |||
XM_005257468.4 | XP_005257525.1 | 7 | 921 | |||
XM_011524922.2 | XP_011523224.1 | 7 | 930 | |||
XM_011524923.2 | XP_011523225.1 | 7 | 927 | |||
XM_011524924.2 | XP_011523226.1 | 7 | 923 | |||
XM_011524925.1 | XP_011523227.1 | 8 | 1044 | |||
XM_011524926.2 | XP_011523228.1 | 8 | 1148 | |||
XM_011524928.2 | XP_011523230.1 | 7 | 1085 | |||
XM_011524929.2 | XP_011523231.1 | 8 | 999 | |||
XM_011524930.1 | XP_011523232.1 | 7 | 721 | |||
XM_011524931.2 | XP_011523233.1 | 6 | 555 | |||
XM_011524933.2 | XP_011523235.1 | 9 | 1105 | |||
XM_011524934.2 | XP_011523236.1 | 7 | 929 | |||
XM_011524935.2 | XP_011523237.1 | 7 | 922 | |||
XM_011524936.1 | XP_011523238.1 | 7 | 721 | |||
XM_017024782.1 | XP_016880271.1 | 8 | 1295 | |||
GenBank mRNA | AK000062.1 | - | 4 | 315 | ||
AK124061.1 | - | 7 | 721 | |||
BC034364.1 | - | 8 | 954 | |||
BC101736.1 | - | 3 | 244 | 8069242 |