Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phyhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013081.1 | NP_001013099.1 | 2 | 148 | ||
XM_006233777.2 | XP_006233839.1 | 3 | 532 | |||
XM_006233778.2 | XP_006233840.1 | 3 | 532 | |||
XM_006233779.2 | XP_006233841.1 | 3 | 1225 | |||
XM_006233780.2 | XP_006233842.1 | 3 | 288 | |||
GenBank mRNA | BC091389.1 | - | 2 | 148 | 7321523 | |
FQ212963.1 | - | 2 | 164 | |||
FQ219402.1 | - | 2 | 169 | |||
FQ223356.1 | - | 2 | 155 |