Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013083.2 | NP_001013101.2 | 7 | 619 | ||
GenBank mRNA | CF249031.1 | - | 4 | 305 | ||
FQ226028.1 | - | 4 | 302 | |||
FQ234275.1 | - | 3 | 168 |