Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhrs7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013098.1 | NP_001013116.1 | 6 | 1233 | ||
GenBank mRNA | AB108671.1 | - | 6 | 1233 | ||
BC104713.1 | - | 6 | 1139 | 7370181 |