Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FTSJ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF327355.1 | - | 4 | 399 | ||
AK307398.1 | - | 4 | 413 | |||
AK315010.1 | - | 5 | 349 | |||
BC036710.2 | - | 4 | 308 | 4641078 |