Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dazap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013107.1 | NP_001013125.1 | 4 | 564 | ||
XM_006242321.2 | XP_006242383.1 | 4 | 959 | |||
XM_006242322.2 | XP_006242384.1 | 5 | 836 | |||
GenBank mRNA | BC062052.1 | - | 4 | 564 | 5598966 |