Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Guk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013115.2 | NP_001013133.2 | 2 | 128 | ||
XM_006246477.2 | XP_006246539.1 | 3 | 265 | |||
XM_006246478.2 | XP_006246540.1 | 2 | 230 | |||
XM_006246479.2 | XP_006246541.1 | 1 | 73 | |||
XM_006246483.1 | XP_006246545.1 | 3 | 180 | |||
XM_008767812.1 | XP_008766034.1 | 3 | 314 | |||
XM_008767813.1 | XP_008766035.1 | 2 | 889 | |||
XM_008767814.1 | XP_008766036.1 | 2 | 138 | |||
XM_008767815.1 | XP_008766037.1 | 2 | 885 | |||
GenBank mRNA | AF354443.1 | - | 2 | 79 | ||
FM065046.1 | - | 2 | 128 |