Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013385.2 | NP_001013403.1 | 8 | 2215 | ||
NM_001291045.1 | NP_001277974.1 | 8 | 2215 | |||
GenBank mRNA | AY255558.1 | - | 1 | 266 | ||
BC072635.1 | - | 11 | 2592 | |||
BC080284.1 | - | 8 | 1986 | 6402528 |
Set Membership: |
G-protein coupled receptor |