Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017653.3 | NP_060123.3 | 3 | 535 | ||
XM_006722488.2 | XP_006722551.1 | 3 | 530 | |||
XM_006722490.2 | XP_006722553.1 | 3 | 530 | |||
XM_006722491.2 | XP_006722554.1 | 3 | 530 | |||
XM_006722492.3 | XP_006722555.1 | 3 | 530 | |||
XM_011526036.1 | XP_011524338.1 | 3 | 530 | |||
XM_011526037.1 | XP_011524339.1 | 3 | 532 | |||
XM_011526038.1 | XP_011524340.1 | 3 | 530 | |||
XM_011526039.1 | XP_011524341.1 | 3 | 530 | |||
XM_011526040.1 | XP_011524342.1 | 3 | 464 | |||
XM_011526041.1 | XP_011524343.1 | 3 | 530 | |||
XM_011526042.1 | XP_011524344.1 | 3 | 530 | |||
XM_017025795.1 | XP_016881284.1 | 3 | 532 | |||
XM_017025796.1 | XP_016881285.1 | 3 | 528 | |||
XM_017025797.1 | XP_016881286.1 | 3 | 468 | |||
XM_017025798.1 | XP_016881287.1 | 3 | 530 | |||
XM_017025799.1 | XP_016881288.1 | 3 | 530 | |||
XM_017025800.1 | XP_016881289.1 | 3 | 530 | |||
XM_017025801.1 | XP_016881290.1 | 3 | 532 | |||
GenBank mRNA | AK074611.1 | - | 3 | 530 | ||
AK091256.1 | - | 4 | 504 | |||
AK291303.1 | - | 3 | 500 | |||
AK296579.1 | - | 3 | 492 | |||
AK315091.1 | - | 4 | 476 | |||
AY364250.1 | - | 2 | 170 | |||
BC064394.1 | - | 3 | 517 | 5752466 |